Would you like to log in again?
Clusterin Rat ELISA, BioVendor
Supplier: BioVendor
RD391034200R
Clusterin is a highly conserved disulfide-linked secreted heterodimeric glycoprotein of 75–80 kDa but truncated forms targeted to the nucleus have also been identified.
  • Calibration Range: 4-128 ng/mL
  • Limit of Detection: 0.7 ng/mL
  • Serum, Urine: 5 uL/well
Clusterin (Apolippoprotein J; SP-40,40; TRPM-2; SGP-2; pADHC-9; CLJ; T64; GP III; XIP8) is a highly conserved disulfide-linked secreted heterodimeric glycoprotein of 75–80 kDa but truncated forms targeted to the nucleus have also been identified. The protein is constitutively secreted by a number of cell types including epithelial and neuronal cells and is a major protein in physiological fluids including plasma, milk, urine, cerebrospinal fluid and semen. Due to its wide breath of tissue distribution many diverse physiological functions have been attributed to Clusterin including sperm maturation, membrane recycling, lipid transportation, tissue remodelling, complement inhibition and cell-cell or cell-substratum interactions. Moreover, it was proposed that Clusterin functions as an extracellular chaperon that stabilizes stressed proteins in a folding-competent state, and that the protein is involved in programmed cell death. Another defining prominent function of Clusterin is its induction in many severe physiological disturbances states including kidney degenerative diseases, prostate and vesicle carcinogenesis, ovarian cancer and several neurodegenerative conditions (Alzheimer’s disease). Recent study demonstrates, that serum Clusterin level increases significantly in diabetic type II patients and in patients with developing coronary heart disease, or myocardial infarction. These data raise the possibility that elevated Clusterin levels in serum may represent a strong indication of vascular damage. In patients with systemic lupus erythematosus (SLE), reduced serum Clusterin levels correlated inversely with disease activity. Lowered Clusterin levels could be involved in the pathogeneses of SLE on account of decreased protective effects. Another interesting observation obtained in rat model suggests that measurement of urinary Clusterin levels may be a useful clinical marker for the severity of renal tubular damage. Furthermore, urinary Clusterin may also help to differentiate between tubular and glomerular forms of proteinuria.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Clusterin Rat ELISA 1 kit RUO RD391034200R 200068-240 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty