Would you like to log in again?
S100G Human ELISA, BioVendor
Supplier: BioVendor
RD191225200R
S100G appear to be involved in neurotrophic and/or neuroprotective processes.
  • Calibration Range: 25-800 pg/mL
  • Limit of Detection: 7.7 pg/mL
  • Plasma (EDTA, citrate, heparin), Serum: 5 uL/well
S100G (S100 calcium binding protein G, Calbindin 3 (CALB3), CaBP9K, CABP1) is a member of the S100 family of calcium-binding proteins. The gene encoding human S100G is located on the X-chromosome (Xq22) and consists of three exons and contains four Alu repeats. The S100G protein (molecular weight 9 kDa) is monomeric and contains two Ca2+ binding motifs, a classical C-terminal EF-hand with a canonical Ca2+ binding loop and an S100-specific N-terminal EF-hand with a modified Ca2+ binding loop called ‘‘pseudo EF-hand’’. S100G protein is present in many organs, cartilage, bone and certain teeth, such as the ameloblasts of incisors and molars. In addition, S100G mRNA is detected in the proximal small intestine, but not in human kidney, uterus or placenta (however, the protein is present in these tissues in other species). S100G is also present in mammalian intestinal epithelial cells (enterocytes). S100G mediates the transport of calcium across the enterocytes from the apical side, where entry is regulated by the calcium channel TRPV6, to the basolateral side, where calcium pumps such as PMCA1 utilize intracellular adenosine triphosphate to pump calcium into the blood. The transport of calcium across the enterocyte cytoplasm appears to be rate-limiting for calcium absorption in the intestine; the presence of calbindin increases the amount of calcium crossing the cell without raising the free concentration. Expression of S100G, like that of calbindin-D28k, is stimulated by the active vitamin D metabolite, calcitriol although the precise mechanisms are still controversial. In mice that lack the receptor for vitamin D, S100G is reduced, but not absent. The members of the S100 family are multifunctional signaling proteins that influence with many cellular events. S100G appear to be involved in neurotrophic and/or neuroprotective processes, but the mechanisms of action are not completely understood. S100G has protective neurotrophic effects during brain development, and alterations in level may serve as an early, quantitative indicator of fetal brain damage in some biological fluids (eg, cord blood). Calcium-regulatory molecule S100G is dynamically expressed and regulated in the uterine endometrium during pregnancy and suggesting that regulation of calcium ion concentration may be critical for the establishment and maintenance of pregnancy in pigs.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
S100G Human ELISA 1 kit RUO RD191225200R 101076-624 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty