S100A11 Human ELISA, Multispecies specificity,, BioVendor
RD191220100R
102552-686EA 908.85 USD
102552-686
S100A11 Human ELISA, Multispecies specificity,, BioVendor
S100A11 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs.
  • Calibration Range: 0.313-20 ng/mL
  • Limit of Detection: 0.01 ng/mL
  • Amniotic fluid, Bronchoalveolar lavage, Serum: (serum) 50 uL/well (BALF) 10 uL/well
S100A11 (S100C; calgizzarin) was first isolated from chicken gizzard smooth muscle. Human homologue was later identified in human colorectal cancer cells and in colorectal normal mucosa, with much higher expression in the cancer cells. S100A11 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. The polypeptide chain consists of 105 amino acid residues giving molecular weight of unmodified monomeric protein 11.74 kDa. Disulphide bridge linking two monomers in the dimer via Cys-11 residues was found in its structure. The protein can be phosphorylated on some residues from which at least phosphorylation on Thr-10 causes significant structural changes. S100A11 binds to annexins A1 and A2, the DNA-dependent ATPase Rad54B, p53 and RAGE. It was proposed that up-regulated chondrocyte expression of S100A11 (as RAGE ligand) in OA cartilage and RAGE signaling through the p38 MAPK pathway promote inflammation-associated chondrocyte hypertrophy. RAGE signaling mediated by S100A11 thereby might have the potential to contribute to the progression of OA. S100A11 has also been shown to be secreted and to exert RAGE dependent signaling in human keratinocytes. S100A11 was found in many different human both normal and cancer tissues. S100A11 appears to play distinct roles depending on the tumour involved. In bladder carcinoma or renal carcinoma expression is related to tumor suppression however in prostate cancer, breast and pancreatic cancer S100A11 is thought to be a tumor promoter. S100A11 has been also indicated as one of potential biomarkers of infective endocarditis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.