Would you like to log in again?
Angiopoietin-Like Protein 3, ANGPTL 3, Human ELISA, BioVendor
Supplier: BioVendor
RD191092200R
ANGPTL3 is structurally similar to angiopoietins, which are vascular endothelial growth factors.
  • Calibration Range: 12.5-400 ng/mL
  • Limit of Detection: 1.08 ng/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 30 uL/well
Angiopoietin-like Protein 3 (Angptl3) is one from six members of Angiopoietin-like (Angptl) family of proteins, which has been identified as orphan ligands with structural similarity to angiopoietins. Angptl3 has 490-amino acid residues and its expression is restricted to the liver. Angptl3 and Angptl4 have been shown to regulate fat, lipid or glucose metabolic homeostasis. Angptl3 is a hepatocyte-derived circulating factor that affect lipid metabolism and is involved in regulating lipid storage and breakdown. Overexpression of Angptl3 or intravenous injection of the purified protein in mice elicited an increase in circulating plasma lipid levels. Angptl3 decreases very low density lipoprotein (VLDL) triglyceride clearance by inhibiting lipoprotein lipase (LPL) activity, directly targeting adipose cells to activate lipolysis. This results in an increased release of free fatty acid (FFA) and glycerol from adipocytes. These observations indicate that Angptl3 might regulate lipid metabolism by inhibiting LPL and by stimulating lipolysis in adipocytes. Recent findings suggest that elevated levels of Angptl3 in diabetic states might be involved in inducing hypertriglyceridemia and hyperfattyacidemia in diabetes and obesity. Hypertriglyceridemia causes triglyceride accumulation in peripheral tissues such as skeletal muscles to enhance insulin- and leptin- resistance, and in vessel walls to induce atherosclerosis. Hyperfattyacidemia also affects the regulation of insulin secretion. Taken together, these findings suggest that abnormalities in the regulation of Angptl3 might be involved in the pathogenesis of metabolic syndrome. Angptl3 also suggest a possible role in the regulation of proliferation of new vessel from preexisting capillaries; a process termed angiogenesis also plays a key role in the progression of solid tumor growth, diabetic retinopathies, psoriasis, inflammation, and rheumathoid arthritis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Angiopoietin-Like Protein 3 (ANGPTL3) Human ELISA 1 kit RUO RD191092200R 102552-072 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty