Would you like to log in again?
VEGF-R3/FLT-4 Human ELISA, BioVendor
Supplier: BioVendor
RAF141R
The sandwich ELISA for detecting and reliable measuring of solubilized VEGFR-3 from tumour tissue, lymphatic vessels and different kind of endothelial cells.
  • Calibration Range: 0.6-40 ng/mL
  • Limit of Detection: 0.03 ng/mL
  • Cell culture supernatant, Plasma-Citrate, Serum: 50 µL (1:10 prediluted)
The main function of the lymphatic system is to return plasma back to the circulating blood. Plasma fluid, macromolecules and cells, such as leucocytes and activated antigen-presenting cells, enter the blind-ended lymphatic vessels. Lymph is then transported toward collecting lymphatic vessels and is returned to the blood circulation. First growth factors and molecular markers specific for lymphatic vessels were discovered only about ten years ago. This is rather astonishing, given the importance of the lymphatic system in maintaining fluid homeostasis and its involvement in the pathogenesis of many diseases, including cancer. Scientific studies over the past years have revealed a signal-transduction system for lymphatic endothelial cell growth, migration and survival. This system is formed by VEGF-C and VEGF-D and their shared receptor VEGFR-3/FLT-4. Homozygous deletion of VEGFR-3 in mice leads to defects in blood-vessel remodelling and embryonic death at mid-gestation, indicating an early vascular function for the receptor. Furthermore mouse studies suggest that upregulation and activation of VEGFR-3 have functional roles in sprouting angiogenesis and that VEGFR-3 is an important effector of the vascular phenotype resulting from Notch inhibition. Heterozygous missense point mutations leading to receptor kinase inactivation have been found in VEGFR-3 of patients with Milroy disease, a rare autosomal dominant lymph oedema. Metastatic tumour spread through the blood or lymphatic vessels in human cancer, often associated with regional lymph-node metastasis, represents the most important prognostic factor for carcinoma patients. Several studies have shown positive correlations between VEGF-C, VEGF-D and VEGFR-3 expression and vascular invasion, lymphatic vessel and lymph node involvement. VEGF-C expression in tumour cells may be induced by growth factors or proinflammatory cytokines. High levels of VEGF-C or VEGF-D also enhance lymphatic metastasis in various experimental models. In some tumours, processed VEGF-C or VEGF-D may be generated, which mainly target VEGFR-3, but also VEGFR-2, which is often upregulated in tumour blood vessels. In lymphoma patients, high expression of VEGFR-3 can been found in lymph endothelial cells isolated from the tumour tissue. In corneal inflammation mouse models, dendritic cells expressing both VEGFR-3 and VEGF-C could be detected, suggesting that immune cells may respond to lymphangiogenic signals and induce lymphangiogenesis. Lymphangiogenesis research has revealed important therapeutic options for human diseases such as lymph oedema and other oedemas that will enter clinical development in the near future. The importance of lymph-node metastasis in the spread of cancer to distant organs needs better understanding before applying the newly acquired knowledge to patients. In this context, the roles of VEGF-C and VEGFR-3 upregulation in tumour angiogenesis need to be explored for additional therapeutical application. The sandwich ELISA for detecting and reliable measuring of solubilized VEGFR-3 from tumour tissue, lymphatic vessels and different kind of endothelial cells will certainly help to understand the mechanisms of lymphatic metastasis including the identification of tumour determinants that are important for the spread of tumour cells.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
VEGF-R3/FLT-4 Human ELISA 1 kit RUO RAF141R 102552-674 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty