Interleukin-6, IL-6, Human ELISA, High Sensitivity, BioVendor
Supplier: BioVendor
RGP013R
102552-440EA 595 USD
102552-440
Interleukin-6, IL-6, Human ELISA, High Sensitivity, BioVendor
Assays ELISAs
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms.
  • Calibration Range: 1.56–50 pg/mL
  • Limit of Detection: 0.8 pg/mL
  • Cell culture supernatant, Plasma, Serum: 100 uL/well
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms. IL-6 is usually not produced constitutively by normal cells, but its expression is readily induced by a variety of cytokines, lipopolysaccharide or viral infections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.