Interleukin-6, IL-6, Monkey ELISA, BioVendor
Supplier: BioVendor
RAF073R
102552-394EA 562.9 USD
102552-394
Interleukin-6, IL-6, Monkey ELISA, BioVendor
Assays ELISAs
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms.
  • Calibration Range: 1.9-120 U/mL
  • Limit of Detection: 1.25 U/mL
  • Baboon serum, Cynomolgus serum: 50 uL/well
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms. IL-6 is usually not produced constitutively by normal cells, but its expression is readily induced by a variety of cytokines, lipopolysaccharide or viral infections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.