Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Interleukin-22, IL-22, Mouse ELISA, BioVendor

Supplier: BioVendor
RAF059R
In the mouse, the IL-22 gene is located on chromosome 10, in the same region as the IFN gamma gene.

  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 1.6 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Serum: 50 uL/well

IL-22/TIF(IL-10-related T cell-derived inducible factor) is a new cytokine originally identified as a gene induced by IL-9 in murine T lymphocytes, and showing 22% amino acid identity with IL-10. In the mouse, the IL-22 gene is located on chromosome 10, in the same region as the IFN gamma gene. Although it is a single copy gene in BALB/c and DBA/2 mice, the IL-22 gene is duplicated in other strains such as C57Bl/6, FVB and 129. The two copies, which show 98% nucleotide identity in the coding region, were named IL-TIF alpha and IL-TIF beta with the IL-TIF beta gene being either differentially regulated, or not expressed at all. IL-22 is produced by activated Th1 and NK cells acting primarily on epithelial cells and is involved in inflammatory responses. Neither resting nor activated immune cells express IL-22 receptor, and IL-22 does not have any effects on these cells in vitro and in vivo. In contrast, cells of the skin and the digestive and respiratory systems represent putative targets of this cytokine. Thus IL-22 does not serve the communication between immune cells but is a T cell mediator that directly promotes the innate, nonspecific immunity of tissues. IL-22 serves as a protective molecule to counteract the destructive nature of the immune response to limit tissue damage. Interleukin-22 (IL-22) is a cytokine that regulates the production of acute phase proteins of the immunological response. On binding to its cognate receptor (IL-22R1), which is associated to the interleukin-10 receptor 2 (IL-10R2), IL-22 promotes activation of signal transducer and activator of transcription (STAT) pathway and several other cellular responses. A soluble receptor termed interleukin-22 binding protein (IL-22BP) is also able to bind to IL-22 as a natural protein antagonist, and probably provides systemic regulation of IL-22 activity. IL-22, in contrast to its relative IFN-gamma, regulates the expression of only a few genes in keratinocytes. This is due to varied signal transduction. The IL-22 effects are transcriptional and either independent of protein synthesis and secretion, or mediated by a secreted protein. Inflammatory conditions, but not keratinocyte differentiation, amplify the IL-22 effects. IL-22 application in mice enhances cutaneous S100A9 and MMP1 expression. Psoriatic patients show strongly elevated IL-22 plasma levels, which correlated with the disease severity. IL-22 plays a protective role in T cell-mediated hepatitis induced by Concanavalin A (Con A), acting as a survival factor for hepatocytes. IL-22 is present in high quantities in the blood of Crohn's disease patients in contrast to IFN-gamma and IL-17.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.




 

ORDER

Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Interleukin-22 (IL-22) Mouse ELISA 1 kit RUO RAF059R 102552-368 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty