Interleukin-2, IL-2, Human ELISA, BioVendor
Supplier: BioVendor
RAF056R
102552-356EA 1054.94 USD
102552-356
Interleukin-2, IL-2, Human ELISA, BioVendor
Assays ELISAs
Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens.
  • Calibration Range: 18.8-1200 pg/mL
  • Limit of Detection: 9.1 pg/mL
  • Amniotic fluid, Cell culture supernatant, Plasma, Serum, Urine, White blood: 50 uL/well
Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens. IL-2 plays a pivotal role in for the expansion of most T-cells, natural killer cells and B-cells during certain phases of their response. IL-2 gene expression is regulated at the transcriptional level by several activation pathways. Antigen-specific proliferation of helper and cytotoxic T-lymphocytes following stimulation is critically dependent on IL-2 expression, secretion, and binding to receptors for IL-2 induced in an autocrine fashion on the surface of T-cells. Apart from its most important role to mediate antigen-specific T-lymphocyte proliferation, IL-2 modulates the expression of IFN-g and major histocompatibility antigens, stimulates proliferation and differentiation of activated B-cells, augments natural killer cell activity and inhibits granulocyte-macrophage colony formation. Alterations in the ability of T-cells to synthesize IL-2 have been observed in physiologic and pathologic states.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.