Would you like to log in again?
Interleukin-1 beta, IL-1b, Mouse ELISA, BioVendor
Supplier: BioVendor
RAF049R
IL-1b has been shown to induce fever and is thought to contribute to wasting of muscles.
  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 1.2 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well
Interleukin-1 (IL-1), originally described in 1972 as lymphocyte activating factor (LAF) for its effects on thymocytes, is a polypeptide cytokine with two molecular forms. Both forms appear to mediate identical ranges of biological activity which include synthesis of the acute phase proteins by hepatocytes, chemotaxis of polymorphonucleocytes, and release of polymorphonucleocytes from blood and bone marrow. These effects coined the acronym leukocyte endogenous mediator (LEM). Early researchers also called IL-1b endogenous pyrogen, and it has been shown to induce fever and is thought to contribute to wasting of muscles (PIF, proteolysis inducing factor). Other activities associated with IL-1 are the induction of Prostaglandin E2 by synovial cells and release of collagenase with resulting destruction of cartilage and bone resorption (catabolin, osteoclast activation factor). In addition, IL-1, has multiple immunological functions including enhancement of IL-2 production by T cells and activation of B-cells (BAF) and thymocytes. A true pleiotrope, IL-1 may have tumoricidal activity via its release of IL-2 and interferon gamma and be indirectly antiviral by stimulating fibroblasts to release interferon beta. In its role as mediator of sepsis, IL-1 has most recently been described as enhancing the growth of virulent E.coli. The two distinct molecular forms of IL-1 are thought to be derived from two genes. After transcription, as 31 kD precursor polypeptide is cleaved to give rise to mostly cell membrane associated IL-1a and secreted IL-1b. Both have the same molecular weight of 15 kD but have different isoelectric points of 5 and 7, respectively. Despite sequence homology of only 20%, both forms are thought to bind to the same receptor. IL-1 inhibitors that vary only in their degree of glycosylation have been described to bind to the IL-1 receptor. These inhibitors are structurally related to IL-1b and may be important in regulation of IL-1b action. Low levels of IL-1b have been reported in normal serum. It is thought that IL-1 genes are induced to respond to tissue damage or in infection. Elevated levels have been reported in a number of infectious disease conditions and in noninfectious inflammatory conditions such as Crohn’s disease. In addition to elevated serum levels, IL-1 has been found in synovial fluids of patients with rheumatoid arthritis and in cerebrospinal fluid after neurological inflammation or insult. At the other end of the spectrum, low levels of IL-1 have been found in malnutrition and advanced neoplasia suggesting perhaps a complex immunological and physiological regulatory role for this cytokine.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Interleukin-1 beta (IL-1b) Mouse ELISA 1 kit RUO RAF049R 102552-350 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty