Interleukin-17A, IL-17A, Human ELISA, BioVendor
Supplier: BioVendor
RAF039R
102552-328EA 1055.05 USD
102552-328
Interleukin-17A, IL-17A, Human ELISA, BioVendor
Assays ELISAs
Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism.
  • Calibration Range: 1.6-100 pg/mL
  • Limit of Detection: 0.5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-Heparin, Serum: 50 uL/well
Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism. IL-17 expression in airways is upregulated upon allergen exposure such as in allergic asthma. Its crucial role in T-cell activation suggests that IL-17 is involved in the development of autoimmune arthritis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.