Interleukin-12 Total, p40 + p70, IL-12 Mouse ELISA, BioVendor
Supplier: BioVendor
RAF033R
102552-314EA 495 USD
102552-314
Interleukin-12 Total, p40 + p70, IL-12 Mouse ELISA, BioVendor
IL-12 is a heterodimeric cytokine which shares the p40 subunit.
  • Calibration Range: 31.3-2000 pg/mL
  • Limit of Detection: 1.9 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well
Cytokines and chemokines and related molecules, Oncology
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.