IFN-gamma Human ELISA, High Sensitivity, BioVendor
Supplier: BioVendor
RGP010R
102552-284EA 595 USD
102552-284
IFN-gamma Human ELISA, High Sensitivity, BioVendor
Assays ELISAs
IFNγ is a homodimeric glycoprotein containing approximately 21 to 24 kD subunits.
  • Calibration Range: 0.78–25 pg/mL
  • Limit of Detection: 0.69 pg/mL
  • Cell culture supernatant, Plasma, Serum: 100 uL/well
IFNγ is a homodimeric glycoprotein containing approximately 21 to 24 kD subunits. Production of IFNg is a function of T cells and NK cells. IFNγ is produced during infection by T cells of the cytotoxic/suppressor phenotype (CD8) and by a subtype of helper T cells, the Th1 cells. IFNγ is a diagnostic tool for distinguishing tuberculous from other nontuberculous ascites. It is important in the design and monitoring of immunotherapy for multiple sclerosis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.