Resistin Human ELISA, BioVendor
Supplier: BioVendor
RD191016100
101215-162EA 735.23 USD
101215-162
Resistin Human ELISA, BioVendor
Assays ELISAs
Human resistin contains 108 amino acids as a prepeptide, and its hydrophobic signal peptide is cleaved before its secretion.
  • Calibration Range: 1-50 ng/mL
  • Limit of Detection: 0.012 ng/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 35 uL/well
Resistin, a product of the RSTN gene, is a peptide hormone belonging to the class of cysteinerich secreted proteins which is termed the RELM family, and is also described as ADSF (Adipose Tissue-Specific Secretory Factor) or FIZZ3 (Found in Inflammatory Zone). Human resistin contains 108 amino acids as a prepeptide, and its hydrophobic signal peptide is cleaved before its secretion. Resistin circulates in human blood as a dimeric protein consisting of two 92 amino acid polypeptides, which are disulfide-linked via Cys26.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.