Anti-BCL9L Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF4967
103244-042EA 663.78 USD
103244-042
Anti-BCL9L Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: BCL9L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR