Anti-IL2RB Mouse Monoclonal Antibody (Phycoerythrin) [clone: 27302]

Supplier: Novus Biologicals
FAB224P
103239-486EA 533.63 USD
103239-486
Anti-IL2RB Mouse Monoclonal Antibody (Phycoerythrin) [clone: 27302]
Antibodies
Type: Primary Antibodies
Antigen: IL2RB
Clonality: Monoclonal
Clone: 27302
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR