Anti-KIR2DL4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 181703]

Supplier: Novus Biologicals
FAB2238A-100
103239-472EA 450.77 USD
103239-472
Anti-KIR2DL4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 181703]
Type: Primary Antibodies
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: 181703
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR