Anti-CCR9 Rat Monoclonal Antibody (Fluorescein) [clone: 242503]

Supplier: Novus Biologicals
FAB2160F-100
103239-434EA 389.02 USD
103239-434
Anti-CCR9 Rat Monoclonal Antibody (Fluorescein) [clone: 242503]
Antibodies
Type: Primary Antibodies
Antigen: CCR9
Clonality: Monoclonal
Clone: 242503
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR