Anti-LYVE1 Rat Monoclonal Antibody (Phycoerythrin) [clone: 223322]

Supplier: Novus Biologicals
FAB2125P
103239-388EA 562.55 USD
103239-388
Anti-LYVE1 Rat Monoclonal Antibody (Phycoerythrin) [clone: 223322]
Antibodies
Type: Primary Antibodies
Antigen: LYVE1
Clonality: Monoclonal
Clone: 223322
Conjugation: Phycoerythrin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR