Anti-CD200R1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 380525]

Supplier: Novus Biologicals
FAB3414A
103239-092EA 562.55 USD
103239-092
Anti-CD200R1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 380525]
Antibodies
Type: Primary Antibodies
Antigen: CD200R1
Clonality: Monoclonal
Clone: 380525
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR