Anti-BSG Mouse Monoclonal Antibody (Phycoerythrin) [clone: TRA-1-85]

Supplier: Novus Biologicals
FAB3195P
103239-032EA 533.63 USD
103239-032
Anti-BSG Mouse Monoclonal Antibody (Phycoerythrin) [clone: TRA-1-85]
Antibodies
Type: Primary Antibodies
Antigen: BSG
Clonality: Monoclonal
Clone: TRA-1-85
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR