Anti-OV-6 Mouse Monoclonal Antibody [clone: OV-6] (PE (Phycoerythrin))

Supplier: Novus Biologicals
FAB2020P
103238-926EA 450.77 USD
103238-926
Anti-OV-6 Mouse Monoclonal Antibody [clone: OV-6] (PE (Phycoerythrin))
Type: Primary Antibodies
Antigen:
Clonality: Monoclonal
Clone: OV-6
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR