Anti-LILRB1 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 292305]

Supplier: Novus Biologicals
FAB20171P-100
103238-908EA 504.71 USD
103238-908
Anti-LILRB1 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 292305]
Antibodies
Type: Primary Antibodies
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292305
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR