Anti-EPHA2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 371805]

Supplier: Novus Biologicals
FAB3035G
103238-796EA 562.55 USD
103238-796
Anti-EPHA2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 371805]
Antibodies
Type: Primary Antibodies
Antigen: EPHA2
Clonality: Monoclonal
Clone: 371805
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR