Anti-SDC1 Rat Monoclonal Antibody (Fluorescein) [clone: 300506]

Supplier: Novus Biologicals
FAB2966F
103238-776EA 267.69 USD
103238-776
Anti-SDC1 Rat Monoclonal Antibody (Fluorescein) [clone: 300506]
Type: Primary Antibodies
Antigen: SDC1
Clonality: Monoclonal
Clone: 300506
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR