Anti-TIMD4 Mouse Monoclonal Antibody (Alexa Fluor 594) [clone: 921832]

Supplier: Novus Biologicals
FAB2929T-025
103238-752EA 258.86 USD
103238-752
Anti-TIMD4 Mouse Monoclonal Antibody (Alexa Fluor 594) [clone: 921832]
Antibodies
Type: Primary Antibodies
Antigen: TIMD4
Clonality: Monoclonal
Clone: 921832
Conjugation: Alexa Fluor 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR