Anti-TIMD4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 921832]

Supplier: Novus Biologicals
FAB2929A-025
103238-744EA 302.25 USD
103238-744
Anti-TIMD4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 921832]
Antibodies
Type: Primary Antibodies
Antigen: TIMD4
Clonality: Monoclonal
Clone: 921832
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR