Anti-CCR10 Rat Monoclonal Antibody (Phycoerythrin) [clone: 248918]

Supplier: Novus Biologicals
FAB2815P-100
103238-690EA 591.48 USD
103238-690
Anti-CCR10 Rat Monoclonal Antibody (Phycoerythrin) [clone: 248918]
Antibodies
Type: Primary Antibodies
Antigen: CCR10
Clonality: Monoclonal
Clone: 248918
Conjugation: Phycoerythrin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR