Anti-KIR2DL3 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 180701]

Supplier: Novus Biologicals
FAB2014A-025
103238-606EA 215.48 USD
103238-606
Anti-KIR2DL3 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 180701]
Antibodies
Type: Primary Antibodies
Antigen: KIR2DL3
Clonality: Monoclonal
Clone: 180701
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR