Anti-TGFB1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 27232]

Supplier: Novus Biologicals
FAB2463A
103238-530EA 620.4 USD
103238-530
Anti-TGFB1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 27232]
Antibodies
Type: Primary Antibodies
Antigen: TGFB1
Clonality: Monoclonal
Clone: 27232
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR