Anti-CD4 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 296712]

Supplier: Novus Biologicals
FAB2410P
103238-462EA 519.17 USD
103238-462
Anti-CD4 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 296712]
Antibodies
Type: Primary Antibodies
Antigen: CD4
Clonality: Monoclonal
Clone: 296712
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR