Anti-FUT4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: MC-480]

Supplier: Novus Biologicals
FAB2155A-100
103238-316EA 577.02 USD
103238-316
Anti-FUT4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: MC-480]
Antibodies
Type: Primary Antibodies
Antigen: FUT4
Clonality: Monoclonal
Clone: MC-480
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR