Anti-CCR6 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 53103]

Supplier: Novus Biologicals
FAB195A-100
103238-202EA 620.4 USD
103238-202
Anti-CCR6 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 53103]
Antibodies
Type: Primary Antibodies
Antigen: CCR6
Clonality: Monoclonal
Clone: 53103
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR