Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL12RB2 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 305719]

Supplier: Novus Biologicals
FAB1959P-025
103238-196
Anti-IL12RB2 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 305719]
Type: Primary Antibodies
Antigen: IL12RB2
Clonality: Monoclonal
Clone: 305719
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty