Anti-CCR10 Rat Monoclonal Antibody (Phycoerythrin) [clone: 314305]

Supplier: Novus Biologicals
FAB3478P-025
103238-076EA 258.86 USD
103238-076
Anti-CCR10 Rat Monoclonal Antibody (Phycoerythrin) [clone: 314305]
Antibodies
Type: Primary Antibodies
Antigen: CCR10
Clonality: Monoclonal
Clone: 314305
Conjugation: Phycoerythrin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR