Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 314305]

Supplier: Novus Biologicals
FAB3478G-100
103238-072EA 634.86 USD
103238-072
Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 314305]
Antibodies
Type: Primary Antibodies
Antigen: CCR10
Clonality: Monoclonal
Clone: 314305
Conjugation: Alexa Fluor 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR