Anti-LILRA1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 586326]

Supplier: Novus Biologicals
FAB30851A
103237-906EA 620.4 USD
103237-906
Anti-LILRA1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 586326]
Antibodies
Type: Primary Antibodies
Antigen: LILRA1
Clonality: Monoclonal
Clone: 586326
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR