Anti-Siglece Rat Monoclonal Antibody (Allophycocyanin) [clone: 750620]

Supplier: Novus Biologicals
FAB5806A
103233-468EA 620.4 USD
103233-468
Anti-Siglece Rat Monoclonal Antibody (Allophycocyanin) [clone: 750620]
Antibodies
Type: Primary Antibodies
Antigen: Siglece
Clonality: Monoclonal
Clone: 750620
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR