Anti-NT5E Mouse Monoclonal Antibody (Phycoerythrin) [clone: 606112]

Supplier: Novus Biologicals
FAB5795P
103233-456EA 533.63 USD
103233-456
Anti-NT5E Mouse Monoclonal Antibody (Phycoerythrin) [clone: 606112]
Antibodies
Type: Primary Antibodies
Antigen: NT5E
Clonality: Monoclonal
Clone: 606112
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Equine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR