Anti-NRP2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 96009]

FAB567G
103233-348EA 493.85 USD
103233-348
Anti-NRP2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 96009]
Type: Primary Antibodies
Antigen: NRP2
Clonality: Monoclonal
Clone: 96009
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR