Anti-TGFBR2 Goat Polyclonal Antibody (Alexa Fluor 700)

Supplier: Novus Biologicals
FAB532N-100
103233-274EA 533.63 USD
103233-274
Anti-TGFBR2 Goat Polyclonal Antibody (Alexa Fluor 700)
Antibodies
Type: Primary Antibodies
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR