Anti-CD24 Mouse Monoclonal Antibody (Allophycocyanin) [clone: ML5]

Supplier: Novus Biologicals
FAB5247A
103233-168EA 533.63 USD
103233-168
Anti-CD24 Mouse Monoclonal Antibody (Allophycocyanin) [clone: ML5]
Antibodies
Type: Primary Antibodies
Antigen: CD24
Clonality: Monoclonal
Clone: ML5
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR