Anti-TNFRSF18 Rat Monoclonal Antibody (Allophycocyanin) [clone: 108619]

Supplier: Novus Biologicals
FAB5241A
103233-148EA 577.02 USD
103233-148
Anti-TNFRSF18 Rat Monoclonal Antibody (Allophycocyanin) [clone: 108619]
Antibodies
Type: Primary Antibodies
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 108619
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR