Anti-EPHB2 Rat Monoclonal Antibody (Allophycocyanin) [clone: 512012]

Supplier: Novus Biologicals
FAB467A
103232-764EA 605.94 USD
103232-764
Anti-EPHB2 Rat Monoclonal Antibody (Allophycocyanin) [clone: 512012]
Antibodies
Type: Primary Antibodies
Antigen: EPHB2
Clonality: Monoclonal
Clone: 512012
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR