Anti-CD47 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 472603]

Supplier: Novus Biologicals
FAB4670A
103232-746EA 562.55 USD
103232-746
Anti-CD47 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 472603]
Antibodies
Type: Primary Antibodies
Antigen: CD47
Clonality: Monoclonal
Clone: 472603
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR