Anti-NAALADL2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 817225]

Supplier: Novus Biologicals
FAB4665G
103232-712EA 466.15 USD
103232-712
Anti-NAALADL2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 817225]
Type: Primary Antibodies
Antigen: NAALADL2
Clonality: Monoclonal
Clone: 817225
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR