Anti-TMEM173 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF6516
103231-472EA 707.17 USD
103231-472
Anti-TMEM173 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: TMEM173
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR