Anti-GPR111 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF6494-SP
103231-446EA 215.48 USD
103231-446
Anti-GPR111 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: GPR111
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR