Anti-UGT1A1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF6490-SP
103231-442EA 215.48 USD
103231-442
Anti-UGT1A1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: UGT1A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR