Anti-KDM4A Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF6434
103231-414EA 663.78 USD
103231-414
Anti-KDM4A Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: KDM4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR